Methyl 4-bromo-2-thiazole-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl-4-bromothiazole-2-carboxylate, Methyl 4-bromothiazole-2-carboxylate

OR302113-1G
APOSOR302113-1GEA 133 EUR
APOSOR302113-1G
Methyl 4-bromo-2-thiazole-carboxylate
Methyl 4-bromo-2-thiazole-carboxylate
Formel: C₅H₄BrNO₂S
Molecular Weight: 222,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11114545
CAS-Nummer: 1025468-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR