1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR302114-1G
APOSOR302114-1GEA 60.9 EUR
APOSOR302114-1G
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid 97%
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,1500 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09743299
CAS-Nummer: 800401-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR