4-Bromo-1-isopropyl-1H-imidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-(prop-2-yl)-1H-imidazole

OR302118-1G
APOSOR302118-1GEA 406 EUR
APOSOR302118-1G
4-Bromo-1-isopropyl-1H-imidazole
4-Bromo-1-isopropyl-1H-imidazole
Formel: C₆H₉BrN₂
Molecular Weight: 189,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22372008
CAS-Nummer: 623577-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR