4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid

Lieferant: Apollo Scientific

OR302119-1G
APOSOR302119-1GEA 220 EUR
APOSOR302119-1G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetic acid
Formel: C₁₁H₁₇BN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16660234
CAS-Nummer: 1083326-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR