5-Bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine
Lieferant: Apollo Scientific
OR302144-1G
APOSOR302144-1GEA
82.8
EUR
APOSOR302144-1G
5-Bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine
5-Bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine
Formel:
C₇H₆BrN₃ Molecular Weight: 212,0500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846319 CAS-Nummer: 885223-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...