5-Bromo-3-isopropyl-1H-indazole

Lieferant: Apollo Scientific

OR302146-1G
APOSOR302146-1GEA 507 EUR
APOSOR302146-1G
5-Bromo-3-isopropyl-1H-indazole
5-Bromo-3-isopropyl-1H-indazole
Formel: C₁₀H₁₁BrN₂
Molecular Weight: 239,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22034709
CAS-Nummer: 1276075-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR