Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95%

Lieferant: Apollo Scientific

OR302149-1G
APOSOR302149-1GEA 835 EUR
APOSOR302149-1G
Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95%
Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide
Formel: C₁₀H₁₃BNO₃·Li
Molecular Weight: 212,967 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16038156
CAS-Nummer: 1014717-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR