3-(3-Furyl)acrylsäure
Lieferant: Apollo Scientific
OR302150-5G
APOSOR302150-5GEA
242
EUR
APOSOR302150-5G
3-(3-Furyl)acrylsäure
3-(3-Furyl)acrylsäure
Formel:
C₇H₆O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075074 CAS-Nummer: 39244-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...