1-(3-Chloropyridin-2-yl)ethanone

Lieferant: Apollo Scientific
Achtung

OR302156-250MG
APOSOR302156-250MGEA 23.2 EUR
APOSOR302156-250MG
1-(3-Chloropyridin-2-yl)ethanone
1-(3-Chloropyridin-2-yl)ethanone
Formel: C₇H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11656222
CAS-Nummer: 131109-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR