2-Hydroxy-3-methylpyrazin
Lieferant: Apollo Scientific
OR302157-1G
APOSOR302157-1GEA
0
EUR
APOSOR302157-1G
2-Hydroxy-3-methylpyrazin
2-Hydroxy-3-methylpyrazine
Formel:
C₅H₆N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646120 CAS-Nummer: 19838-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...