3-Methyldihydro-2H-pyran-4(3H)-one
Lieferant: Apollo Scientific
Synonyme:
3-Methyltetrahydropyran-4-one
OR302159-1G
APOSOR302159-1GEA
393
EUR
APOSOR302159-1G
3-Methyldihydro-2H-pyran-4(3H)-one
3-Methyldihydro-2H-pyran-4(3H)-one
Formel:
C6H10O2 Molecular Weight: 114,1424 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193744 CAS-Nummer: 119124-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...