2-Bromo-N-methylpyridin-4-amine

Lieferant: Apollo Scientific

OR302166-1G
APOSOR302166-1GEA 426 EUR
APOSOR302166-1G
2-Bromo-N-methylpyridin-4-amine
2-Bromo-N-methylpyridin-4-amine
Formel: C₆H₇BrN₂
Molecular Weight: 187,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18259179
CAS-Nummer: 847799-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR