6-Methoxy-1,3-benzothiazol-2-carbonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methoxy-2-benzothiazolecarbonitrile, 6-Methoxybenzo[d]thiazole-2-carbonitrile
OR302169-5G
APOSOR302169-5GEA
95.7
EUR
APOSOR302169-5G
6-Methoxy-1,3-benzothiazol-2-carbonitril
2-Cyano-6-methoxybenzothiazole
Formel:
C₉H₆N₂OS Schmelzpunkt: 129…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010537 CAS-Nummer: 943-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...