2-Bromo-6-methoxybenzothiazole

Lieferant: Apollo Scientific

OR302170-5G
APOSOR302170-5GEA 58 EUR
APOSOR302170-5G
2-Bromo-6-methoxybenzothiazole
2-Bromo-6-methoxybenzothiazole
Formel: C₈H₆BrNOS
Lagertemperatur: Raumtemperatur
MDL: MFCD08459026
CAS-Nummer: 2941-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR