2-Hydroxy-5-nitro-6-picolin
Lieferant: Apollo Scientific
OR302172-5G
APOSOR302172-5GEA
60.9
EUR
APOSOR302172-5G
2-Hydroxy-5-nitro-6-picolin
2-Hydroxy-5-nitro-6-picolin
Formel:
C₆H₆N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092010 CAS-Nummer: 28489-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...