1,6-Dichloro-2,7-naphthyridine

Lieferant: Apollo Scientific
Achtung

OR302178-1G
APOSOR302178-1GEA 0 EUR
APOSOR302178-1G
1,6-Dichloro-2,7-naphthyridine
1,6-Dichloro-2,7-naphthyridine
Formel: C₈H₄Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD26403990
CAS-Nummer: 1335053-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR