2-Hydroxyquinoline-5-carboxylic acid

Lieferant: Apollo Scientific

OR302189-1G
APOSOR302189-1GEA 357 EUR
APOSOR302189-1G
2-Hydroxyquinoline-5-carboxylic acid
2-Hydroxyquinoline-5-carboxylic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11557267
CAS-Nummer: 83734-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR