2-(3-Amino-1H-pyrazol-5-yl)acetic acid

Lieferant: Apollo Scientific

Synonyme: 5-Amino-1H-pyrazole-3-acetic acid

OR302195-1G
APOSOR302195-1GEA 212 EUR
APOSOR302195-1G
2-(3-Amino-1H-pyrazol-5-yl)acetic acid
2-(3-Amino-1H-pyrazol-5-yl)acetic acid
Formel: C₅H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09991896
CAS-Nummer: 174891-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR