3-Formyl-4-methoxypyridin

Lieferant: Apollo Scientific

Synonyme: 4-Methoxypyridine-3-carboxaldehyde, 4-Methoxynicotinaldehyde

OR302197-1G
APOSOR302197-1GEA 226 EUR
APOSOR302197-1G
3-Formyl-4-methoxypyridin
3-Formyl-4-methoxypyridin
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112505
CAS-Nummer: 82257-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR