2-(5-Bromopyridin-2-yl)propan-2-ol
Lieferant: Apollo Scientific
|
Gefahr
|
OR302203-1G
APOSOR302203-1GEA
125
EUR
APOSOR302203-1G
2-(5-Bromopyridin-2-yl)propan-2-ol
2-(5-Bromopyridin-2-yl)propan-2-ol
Formel:
C₈H₁₀BrNO Lagertemperatur: Kühlschrank |
MDL:
MFCD11865230 CAS-Nummer: 290307-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...