2'-Nitroacetanilid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2-Nitrophenyl)acetamid
OR30221-25G
APOSOR30221-25GEA
65.4
EUR
APOSOR30221-25G
2'-Nitroacetanilid
2'-Nitroacetanilid
Formel:
C₈H₈N₂O₃ Molecular Weight: 180,16 g/mol Schmelzpunkt: 90…93 °C Dichte: 1,42 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016991 CAS-Nummer: 552-32-9 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...