Methyl 2-((tert-butoxycarbonyl)amino)isonicotinate

Lieferant: Apollo Scientific

Synonyme: 2-(2,2-Dimethylpropionylamino)isonicotinic acid methyl ester

OR302224-5G
APOSOR302224-5GEA 237 EUR
APOSOR302224-5G
Methyl 2-((tert-butoxycarbonyl)amino)isonicotinate
Methyl 2-((tert-butoxycarbonyl)amino)isonicotinate
Formel: C₁₂H₁₆N₂O₄
Molecular Weight: 252,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11504796
CAS-Nummer: 639091-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR