tert-Butyl (4-formylpyridin-2-yl)carbamate

Lieferant: Apollo Scientific

OR302225-1G
APOSOR302225-1GEA 179 EUR
APOSOR302225-1G
tert-Butyl (4-formylpyridin-2-yl)carbamate
tert-Butyl (4-formylpyridin-2-yl)carbamate
Formel: C₁₁H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09757496
CAS-Nummer: 304873-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR