Methyl-5-methoxynicotinat
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-methoxypyridine-3-carboxylate, Methyl 5-methoxynicotinate
OR302228-1G
APOSOR302228-1GEA
91.5
EUR
APOSOR302228-1G
Methyl-5-methoxynicotinat
Methyl-5-methoxynicotinat
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09265109 CAS-Nummer: 29681-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...