2-(5-Chloropyridin-2-yl)acetonitrile

Lieferant: Apollo Scientific

OR302231-1G
APOSOR302231-1GEA 272 EUR
APOSOR302231-1G
2-(5-Chloropyridin-2-yl)acetonitrile
2-(5-Chloropyridin-2-yl)acetonitrile
Formel: C₇H₅ClN₂
Molecular Weight: 152,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09923938
CAS-Nummer: 185315-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR