1-(Pyridin-2-yl)propan-2-one
Lieferant: Apollo Scientific
|
Achtung
|
OR302235-1G
APOSOR302235-1GEA
77
EUR
APOSOR302235-1G
1-(Pyridin-2-yl)propan-2-one
1-(Pyridin-2-yl)propan-2-one
Formel:
C₈H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092717 CAS-Nummer: 6302-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...