Methyl 3-(1H-pyrrol-2-yl)propanoate

Lieferant: Apollo Scientific
Achtung

OR302239-1G
APOSOR302239-1GEA 71.2 EUR
APOSOR302239-1G
Methyl 3-(1H-pyrrol-2-yl)propanoate
Methyl 3-(1H-pyrrol-2-yl)propanoate
Formel: C₈H₁₁NO₂
Molecular Weight: 153,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09841003
CAS-Nummer: 69917-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR