Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate

Lieferant: Apollo Scientific

OR302241-1G
APOSOR302241-1GEA 464 EUR
APOSOR302241-1G
Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25973573
CAS-Nummer: 1335053-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR