Ethyl-2,7-dimethylimidazo[1,2-a]pyridin-3-carboxylat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate
OR302245-1G
APOSOR302245-1GEA
125
EUR
APOSOR302245-1G
Ethyl-2,7-dimethylimidazo[1,2-a]pyridin-3-carboxylat
Ethyl-2,7-dimethylimidazo[1,2-a]pyridin-3-carboxylat
Formel:
C₁₂H₁₄N₂O₂ Molecular Weight: 218,2500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139510 CAS-Nummer: 81448-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...