Ethyl-4,6-dichloroquinoline-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 4,6-dichloroquinoline-3-carboxylate

OR302253-1G
APOSOR302253-1GEA 63.8 EUR
APOSOR302253-1G
Ethyl-4,6-dichloroquinoline-3-carboxylate
Ethyl-4,6-dichloroquinoline-3-carboxylate
Formel: C₁₂H₉Cl₂NO₂
Molecular Weight: 270,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173348
CAS-Nummer: 21168-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR