2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 95%

Lieferant: Apollo Scientific

Synonyme: 2-((6-Chloro-2-methylpyrimidin-4-yl)amino)-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide, N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide

OR302262-25G
APOSOR302262-25GEA 69.6 EUR
APOSOR302262-25G
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 95%
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formel: C₁₆H₁₃Cl₂N₅OS
Molecular Weight: 394.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11045075
CAS-Nummer: 302964-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR