4-Bromophthalimide

Lieferant: Apollo Scientific

Synonyme: 5-Bromoisoindoline-1,3-dione, 5-bromo-2,3-dihydro-1H-isoindole-1,3-dione

OR302268-100G
APOSOR302268-100GEA 157 EUR
APOSOR302268-100G
4-Bromophthalimide
4-Bromophthalimide
Formel: C₈H₄BrNO₂
Molecular Weight: 226,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466049
CAS-Nummer: 6941-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR