N,N-Diboc-4-amino-6-chloropyrimidine

Lieferant: Apollo Scientific

Synonyme: N,N-DiBoc-4-amino-6-chloropyrimidine, Di-tert-butyl (6-chloropyrimidin-4-yl)imidodicarbonate

OR302269-1G
APOSOR302269-1GEA 102 EUR
APOSOR302269-1G
N,N-Diboc-4-amino-6-chloropyrimidine
N,N-Diboc-4-amino-6-chloropyrimidine
Formel: C₁₄H₂₀ClN₃O₄
Molecular Weight: 329,7800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566080
CAS-Nummer: 354112-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR