4-(2-Pyrrolidin-1-ylethoxy)phenylamine

Lieferant: Apollo Scientific

Synonyme: 4-(2-(Pyrrolidin-1-yl)ethoxy)aniline

OR302272-1G
APOSOR302272-1GEA 194 EUR
APOSOR302272-1G
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07790198
CAS-Nummer: 50609-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR