Quinoline-4,7-diol

Lieferant: Apollo Scientific

OR302275-1G
APOSOR302275-1GEA 279 EUR
APOSOR302275-1G
Quinoline-4,7-diol
Quinoline-4,7-diol
Formel: C₉H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09260461
CAS-Nummer: 955938-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR