But-3-en-1-amine, N-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Boc-amino)-3-butene

OR302285-1G
APOSOR302285-1GEA 114 EUR
APOSOR302285-1G
But-3-en-1-amine, N-BOC protected
But-3-en-1-amine, N-BOC protected
Formel: C₉H₁₇NO₂
Dichte: 0,9293 g/cm³ (25 °C)
Flammpunkt: 101 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11975862
CAS-Nummer: 156731-40-7
UN: 3082
ADR: 9+EHS,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR