2-Chlor-4-iodtoluol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-4-iodo-1-methylbenzene, 1-Chloro-5-iodo-2-methylbenzene
OR30229-100G
OR30229-25G
APOSOR30229-100GEA
78.3
EUR
APOSOR30229-100G
APOSOR30229-25G
2-Chlor-4-iodtoluol
2-Chlor-4-iodtoluol
Formel:
C₇H₆ClI Molecular Weight: 252.48 g/mol Siedepunkt: 118…120 °C/15mmHg Dichte: 1.8 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040872 CAS-Nummer: 83846-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...