(3R,4S)-tert-Butyl 3,4-dihydroxypyrrolidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: rel-(3R,4S)-tert-Butyl 3,4-dihydroxypyrrolidine-1-carboxylate

OR302297-1G
APOSOR302297-1GEA 277 EUR
APOSOR302297-1G
(3R,4S)-tert-Butyl 3,4-dihydroxypyrrolidine-1-carboxylate
(3R,4S)-tert-Butyl 3,4-dihydroxypyrrolidine-1-carboxylate
Formel: C₉H₁₇NO₄
Molecular Weight: 203,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21919047
CAS-Nummer: 186393-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR