5-Bromo-N,N,4-trimethyl-2-pyridinamine

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-N,N,4-trimethylpyridin-2-amine

OR302299-1G
APOSOR302299-1GEA 386 EUR
APOSOR302299-1G
5-Bromo-N,N,4-trimethyl-2-pyridinamine
5-Bromo-N,N,4-trimethyl-2-pyridinamine
Formel: C₈H₁₁BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08272097
CAS-Nummer: 764651-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR