7-Methylpyrazolo[1,5-a][1,3,5]triazin-4(3H)-one

Lieferant: Apollo Scientific

OR302301-1G
APOSOR302301-1GEA 404 EUR
APOSOR302301-1G
7-Methylpyrazolo[1,5-a][1,3,5]triazin-4(3H)-one
7-Methylpyrazolo[1,5-a][1,3,5]triazin-4(3H)-one
Formel: C₆H₆N₄O
Molecular Weight: 150,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12755918
CAS-Nummer: 55457-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR