Benzyl 4-(sulphamoylmethyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

OR302309-1G
APOSOR302309-1GEA 144 EUR
APOSOR302309-1G
Benzyl 4-(sulphamoylmethyl)piperidine-1-carboxylate
Benzyl 4-(sulphamoylmethyl)piperidine-1-carboxylate
Formel: C₁₄H₂₀N₂O₄S
Molecular Weight: 312,385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27501042
CAS-Nummer: 1585225-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR