4-(Azetidin-3-yl)morpholine hydrochloride

Lieferant: Apollo Scientific

OR302316-1G
APOSOR302316-1GEA 0 EUR
APOSOR302316-1G
4-(Azetidin-3-yl)morpholine hydrochloride
4-(Azetidin-3-yl)morpholine hydrochloride
Formel: C₇H₁₄N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11865371
CAS-Nummer: 223381-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR