4-(Azetidin-3-yl)-1-methylpiperazin-2-one dihydrochloride

Lieferant: Apollo Scientific

OR302318-1G
APOSOR302318-1GEA 481 EUR
APOSOR302318-1G
4-(Azetidin-3-yl)-1-methylpiperazin-2-one dihydrochloride
4-(Azetidin-3-yl)-1-methylpiperazin-2-one dihydrochloride
Formel: C₈H₁₅N₃O·2ClH
Molecular Weight: 242,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22581554
CAS-Nummer: 1403766-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR