1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazole-4-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]propan-2-ol, 2-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)propan-2-ol, 2-[1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR302325-1G
APOSOR302325-1GEA 557 EUR
APOSOR302325-1G
1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazole-4-boronic acid, pinacol ester
1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazole-4-boronic acid, pinacol ester
Formel: C₁₃H₂₃BN₂O₃
Molecular Weight: 266,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22378011
CAS-Nummer: 1082503-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR