1-(Pyrimidin-2-yl)-1H-pyrazol-4-amine 96%
Lieferant: Apollo Scientific
OR302329-1G
APOSOR302329-1GEA
679
EUR
APOSOR302329-1G
1-(Pyrimidin-2-yl)-1H-pyrazol-4-amine 96%
1-(Pyrimidin-2-yl)-1H-pyrazol-4-amine
Formel:
C₇H₇N₅ Molecular Weight: 161,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12192741 CAS-Nummer: 1156601-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...