2,2'-Bipyridin-4,4'-diamin
Lieferant: Apollo Scientific
Synonyme:
[2,2'-Bipyridine]-4,4'-diamine, 4,4'-Diamino-2,2'-bipyridine
OR302331-1G
APOSOR302331-1GEA
102
EUR
APOSOR302331-1G
2,2'-Bipyridin-4,4'-diamin
2,2'-Bipyridin-4,4'-diamin
Formel:
C₁₀H₁₀N₄ Molecular Weight: 186,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704188 CAS-Nummer: 18511-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...