4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR302339-250MG
APOSOR302339-250MGEA
104
EUR
APOSOR302339-250MG
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine 95%
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine
Formel:
C₇H₄Cl₂N₂ Schmelzpunkt: 252…254 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038480 CAS-Nummer: 67139-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...