6-Bromopyridine-3,4-diamine

Lieferant: Apollo Scientific

OR302354-1G
APOSOR302354-1GEA 493 EUR
APOSOR302354-1G
6-Bromopyridine-3,4-diamine
6-Bromopyridine-3,4-diamine
Formel: C₅H₆BrN₃
Molecular Weight: 188,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100655
CAS-Nummer: 1033203-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR