4,6-Pyrimidindiamin
Lieferant: Apollo Scientific
Synonyme:
4,6-Pyrimidinediamine
OR302359-1G
APOSOR302359-1GEA
50.9
EUR
APOSOR302359-1G
4,6-Pyrimidindiamin
4,6-Pyrimidindiamin
Formel:
C₄H₆N₄ Molecular Weight: 110,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044572 CAS-Nummer: 2434-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...