5-Aminoisoquinolin-1(2H)-one

Lieferant: Apollo Scientific

Synonyme: 5-Aminoisoquinolin-1-ol, 5-AIQ

OR302365-1G
APOSOR302365-1GEA 411 EUR
APOSOR302365-1G
5-Aminoisoquinolin-1(2H)-one
5-Aminoisoquinolin-1(2H)-one
Formel: C₉H₈N₂O
Molecular Weight: 160.1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06198896
CAS-Nummer: 93117-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR